Learn More
Leica Biosystems Novolink MaxDAB(Polymer)(1250)
Supplier:  Leica Biosystems RE7270CE
Encompass
“Novolink Polymer Detection Systems are used for the visualization of mouse IgG, mouse IgM and rabbit IgG primary antibodies. Novolink Polymer contains component reagents of these systems. The Novolink Polymer Detection Systems utilize a novel controlled polymerization technology to prepare polymeric HRP-linker antibody conjugates. Therefore, the problem of non-specific staining that can occur with Streptavidin/Biotin detection systems due to endogenous biotin does not occur. These products are used in an immunohistochemical (IHC) procedure, which allows the qualitative identification by light microscopy of antigens in sections of formalin-fixed, paraffin-embedded tissue. Post Primary (Rabbit anti-Mouse IgG) is used to detect mouse antibodies. The Novolink Polymer (anti-rabbit Poly-HRP-IgG) recognizes rabbit immunoglobulins, detecting the post primary and any tissue-bound rabbit primary antibodies. Novolink (Polymer), RE72-K, is a two part ready-to-use kit comprising 25mL of No
Catalog No. 50-255-1982
 Restricted Item