Learn More
Leica Biosystems NOVOLINK POLYMER DETECTION KIT
Catalog No. NC1488442
Encompass
The Novolink Polymer Detection Systems utilize a novel Compact Polymer technology. Therefore, the problem of non-specific staining that can occur with Streptavidin/Biotin detection systems due to endogenous biotin does not occur. Novolink Polymer Detection Systems contain pre-diluted, reagents in color coded bottles for ease of use and ultimate convenience.These systems can be used for the visualization of mouse IgG, mouse IgM and rabbit IgG primary antibodies.These detection systems contain Peroxidase Block, Protein Block, Post Primary Block, Novolink Polymer, DAB Chromogen, Novolink DAB Substrate Buffer (Polymer) and Hematoxylin.
Catalog No. NC1488442 Supplier Leica Biosystems Supplier No. RE7150CE
The Novolink Polymer Detection Systems utilize a novel Compact Polymer technology. Therefore, the problem of non-specific staining that can occur with Streptavidin/Biotin detection systems due to endogenous biotin does not occur. Novolink Polymer Detection Systems contain pre-diluted, reagents in color coded bottles for ease of use and ultimate convenience.These systems can be used for the visualization of mouse IgG, mouse IgM and rabbit IgG primary antibodies.These detection systems contain Peroxidase Block, Protein Block, Post Primary Block, Novolink Polymer, DAB Chromogen, Novolink DAB Substrate Buffer (Polymer) and Hematoxylin.