Learn More
MONOD BIO INC NovoLISA Bundle Kit: LuxLink Conjugation Kit and LuxSit Pro Assay Kit
Catalog No. NC4045333
Encompass_Preferred
The NovoLISA bundle kit includes both LuxLink NovoLISA conjugation kit and the 1000 reaction LuxSit Pro Assay Kit. LuxLink NovoLISA conjugation kit is a rapid, amine-reactive kit with split LuxSit™ luciferase fragments (Bit and aLux) for generating antibody conjugates for NovoLISA™ no-wash immunoassays. supports up to 200 μg of antibody. Includes: Bit-TFP ester, tetrazine-aLux, and NHS-TCO linker. the LuxSit™ Pro Kit enables scientists to read out assays for the custom NovoLISA luminescence assays for 1000 reaction. Each NovoLISA Bundle Kit includes (1) assay reaction buffer, (2) substrate, and (3) substrate reconstitution buffer (4) Bit-TFP Ester (5) Tetrazine-aLux, (6) and NHS-TCO
Catalog No. NC4045333 Supplier MONOD BIO INC Supplier No. NLB0101
The NovoLISA bundle kit includes both LuxLink NovoLISA conjugation kit and the 1000 reaction LuxSit Pro Assay Kit. LuxLink NovoLISA conjugation kit is a rapid, amine-reactive kit with split LuxSit™ luciferase fragments (Bit and aLux) for generating antibody conjugates for NovoLISA™ no-wash immunoassays. supports up to 200 μg of antibody. Includes: Bit-TFP ester, tetrazine-aLux, and NHS-TCO linker. the LuxSit™ Pro Kit enables scientists to read out assays for the custom NovoLISA luminescence assays for 1000 reaction. Each NovoLISA Bundle Kit includes (1) assay reaction buffer, (2) substrate, and (3) substrate reconstitution buffer (4) Bit-TFP Ester (5) Tetrazine-aLux, (6) and NHS-TCO