Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies NOVORINSE SOLN 5X 500 ML

Catalog No. NC3364179
Encompass

  • Concentration: 5X
  • Volume: 500 mL
  • Product type: Rinse solution
  • Application: Laboratory instrument rinsing, contaminant removal, system maintenance
  • Instrument compatibility: Liquid chromatography systems, mass spectrometry systems, HPLC, LC-MS systems, autosamplers, injector components
  • Formulation: Concentrated liquid
  • Benefit: Reduces carryover
  • Benefit: Minimizes cross-contamination
  • Benefit: Enhances system longevity
  • Benefit: Improves analytical reliability
  • Compatibility: Agilent workflows
  • Usage: Dilutable for ready-to-use rinse

Catalog No. NC3364179 Supplier Agilent Technologies Supplier No. 872B610
May include imposed supplier surcharges.
Add to Cart
Add to Cart

  • Concentration: 5X
  • Volume: 500 mL
  • Product type: Rinse solution
  • Application: Laboratory instrument rinsing, contaminant removal, system maintenance
  • Instrument compatibility: Liquid chromatography systems, mass spectrometry systems, HPLC, LC-MS systems, autosamplers, injector components
  • Formulation: Concentrated liquid
  • Benefit: Reduces carryover
  • Benefit: Minimizes cross-contamination
  • Benefit: Enhances system longevity
  • Benefit: Improves analytical reliability
  • Compatibility: Agilent workflows
  • Usage: Dilutable for ready-to-use rinse

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.