Learn More
Novus Biologicals™ DOG1/TMEM16A Antibody Blocking Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 55107 |
| Species | Human |
| Purity | >85% |
| Content And Storage | Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.) |
| Formulation | PBS, BSA 0,1 % |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP2-29662. This Peptide Blocks Completely The Immuno Reactivity of The Antibody NBP2-29662 in. Western Blot or Immunohistochemistry by Incubating The Peptide with Equivalent Volume of Antibody for 30 Min (at 37 Degrees C. For Further Blocking Peptide Related Protocol, Click Here .) |
| Gene Alias | ANO1, anoctamin 1, DOG1, ORAOV2, TAOS2, TMEM16A |
| Gene Symbol | ANO1 |
| Molecular Weight (g/mol) | M.W. (theoretical): 0.81 kDa |
| Product Type | DOG1/TMEM16A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.