Learn More
Novus Biologicals™ Histone H3 [Trimethyl Lys18] Antibody Blocking Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 126961 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Lyophilized. Reconstitute in 40 ul of diH20. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB21-1143. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | H3, H3 histone, family 3 A, H3.3 A, H3.3 B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 |
| Gene Symbol | HIST2H3C |
| Product Type | Histone H3 |
| Quantity | 0.04 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.