Learn More
Novus Biologicals™ HMGB1/HMG-1 Antibody Blocking Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 3146 |
| Species | Mouse |
| Accession Number | P09429 |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-2322. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein |
| Gene Symbol | HMGB1 |
| Product Type | HMGB1/HMG-1 |
| Quantity | 0.05 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.