Learn More
Novus Biologicals™ KA1/GRIK4/Glutamate Receptor KA1 Antibody Blocking Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 2900 |
| Species | Mouse |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-76851. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | EAA1, Excitatory amino acid receptor 1, glutamate receptor KA1, Glutamate receptor KA-1, glutamate receptor, ionotropic kainate 4, glutamate receptor, ionotropic, kainate 4, KA1GRIK |
| Gene Symbol | GRIK4 |
| Product Type | KA1/GRIK4/Glutamate Receptor KA1 |
| Quantity | 0.05 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.