Learn More
Novus Biologicals™ LDLR Antibody Blocking Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 3949 |
| Species | Human |
| Purification Method | Peptide affinity purified |
| Purity | Immunogen affinity purified |
| Accession Number | P01130 |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | PBS, 30% glycerol |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-06709. This Peptide is Useful as a Negative Control for Western Blot. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 |
| Gene Symbol | LDLR |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.