Learn More
Novus Biologicals™ ND OLLAS/DYKDDDDK Vector

Description
The pCMV-ND-OLLAS vector provides a convenient vector to express recombinant proteins under control of the CMV promoter with both an N-terminal OLLAS tag and a DYKDDDDK tag, protein will be expressed in the cytoplasm. Insert protein of interest into the vector in frame using the Xba I or EcoRI/Not I restriction sites. Bacterial selection can be performed using Kanamycin, while Neomycin resistance can be used for screening mammalian transfectants.
Recommended Storage
Store at -20°.Avoid freeze-thaw cycles.
Specifications
Specifications
| Storage Buffer | 5mM Tris pH 8 |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 0.01 mg |
| Vector | ND OLLAS/DYKDDDDK Vector |
| For Use With (Application) | For intracellular expression of N-terminally FLAG(DYKDDDDK) and OLLAS dual tandem tagged protein |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.