Learn More
Novus Biologicals™ OMgp Antibody Blocking Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 4974 |
| Species | Human |
| Purity | >50%, by HPLC |
| Accession Number | NP_002535.3 |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulation | Lyophilized from a 5 pmol/ul solution in 5% (v/v) acetonitrile/H2O and TFA salt. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-2557. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | oligodendrocyte myelin glycoprotein, OMGPoligodendrocyte-myelin glycoprotein |
| Gene Symbol | OMG |
| Product Type | OMgp |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.