Learn More
Novus Biologicals™ p19ARF/CDKN2A Antibody Blocking Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 12578 |
| Species | Human |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB 200-106. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | alternative reading frame, ARF, ARF-INK4a, CDK4I, CDKN2A, Cyclin-dependent kinase 4 inhibitor A, cyclin-dependent kinase inhibitor 2 A, cyclin-dependent kinase inhibitor 2 A (p16, inhibits CDK4), cyclin-dependent kinase inhibitor 2 A, isoform 3, cyclin-dependent kinase inhibitor 2 A, isoforms 1/2, cyclin-dependent kinase inhibitor protein, INK4, INK4a, Ink4a/Arf, INK4a-ARF, mitochondrial smARF', MTS1, p14, p14ARF, P16, p16(INK4a), p16-INK4, p16INK4a, p16-INK4a, P19, p19 |
| Gene Symbol | Cdkn2a |
| Product Type | p19ARF |
| Quantity | 0.05 mg |
| Regulatory Status | RUO |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.