Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ pCLNCX Retrovirus Expression Vector
SDP

Catalog No. NBP229502
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229502 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229502 Supplier Novus Biologicals™ Supplier No. NBP229502

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Applications: Retroviral Production

The pCLNCX expression vector is a part of the RetroMax expression system and has been designed to maximize recombinant-retrovirus titers in a simple, efficient, and flexible experimental system. All members of the RetroMax expression vector family (pCLXSN, pCLNCX, pCLNRX, and pCLNDX) have an extended packaging signal (omega+) and are derived from safety modified retrovirus vectors in which the gag open reading frame has been stopped by a point mutation, thereby minimizing the opportunity for replication competent retrovirus production by recombination with packaging genome.

  • RetroMax system takes advantages of two properties of 293 cells (high level of transfectibility and strong E1A-mediated stimulation of CMV promoter controlled transcription)
  • 293 cells of nonmurine origin avoid problem of selective packaging and transfer of VL30 genomes (present in all murine packaging cells)
  • Vector supernatants are free of helper virus and are of sufficiently high titer within 2 days of transient transfection in 293 cells
  • Buffer: 10μg in 20μL 1x TE (10mM Tris, pH 7.5, 1mM EDTA)

Recommended Storage

Store at -20°C. Avoid freeze-thaw cycles.

Specifications

Storage Buffer 10μg in 20μL 1x TE (10mM Tris, pH 7.5, 1mM EDTA)
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 10 μg
Vector pCLNCX Retrovirus Expression Vector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.