Learn More
Novus Biologicals™ Small Intestine Duodenum Tissue Slides (Normal)

Specifications
Specifications
| Cross Reactivity | Human |
| Type | Small Intestine Duodenum Tissue Slides (Normal) |
| For Use With (Application) | Immunohistochemistry-Paraffin |
| Regulatory Status | RUO |
| Species | Human |
| Tissue Condition | Normal |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.