Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ WDR74 Antibody Blocking Peptide
SDP

Catalog No. NBP177140E
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177140E 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177140E Supplier Novus Biologicals™ Supplier No. NBP177140PEP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide containing 18 amino acids near the carboxy terminus of human WDR74. Source: Synthetic (Accession #: Q6RFH5) The WDR74 Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 54663
Species Human
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-77140. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias FLJ10439, FLJ21730, NOP seven-associated protein 1, NSA1, WD repeat domain 74, WD repeat-containing protein 74
Gene Symbol WDR74
Product Type WDR74
Quantity 0.05 mg
Regulatory Status RUO
Source Synthetic
Termination 18 amino acids near the carboxy terminus of human NSA1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.