Learn More
Novus Biologicals™ Chromata ChIP Histone H3 [ac Lys9, ac Lys14] Kit (ChIP)

Description
Includes
10x glycine, IP dilution buffer, H3 K4Me3 positive control antibody, Protein A/G magnetic beads, IP wash buffer 1, IP wash buffer 2, IP wash buffer 3, IP wash buffer 4, IP elution buffer, 5M NaCL, Proteinase K, Purification columns, Purification column Tubes, DNA binding buffer, DNA wash buffer, DNA elution buffer, Human RPL30 positive control primer pair
Kit contains all the necessary components to perform 25 Immunoprecipitation experiments including a positive control antibody. This ChIP kit is specific for Histone H3 acetylated Lysine 9 / Lysine 14.
Specifications
Specifications
| Kit Contents | 10x glycine, IP dilution buffer, H3 K4Me3 positive control antibody, Protein A/G magnetic beads, IP wash buffer 1, IP wash buffer 2, IP wash buffer 3, IP wash buffer 4, IP elution buffer, 5M NaCL, Proteinase K, Purification columns, Purification column Tubes, DNA binding buffer, DNA wash buffer, DNA elution buffer, Human RPL30 positive control primer pair |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Quantity | 25 Immunoprecipitations |
| For Use With (Application) | Chromatin Immunoprecipitation of DNA from fixed Hela cells alongside a no antibody (No Ab) control |
| Target | Chromata ChIP Histone H3 |
| Type | ChIP Histone H3 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.