Learn More
Novus Biologicals™ Chromata ChIP Kit (ChIP)

Description
Includes
RPL30 primer, 10x glycine, IP dilution buffer, H3 K4Me3 positive control antibody, Protein A/G magnetic beads, IP wash buffer 1, IP wash buffer 2, IP wash buffer 3, IP wash buffer 4, IP elution buffer, 5M NaCL, Proteinase K, Purification columns, Purification column Tubes, DNA binding buffer, DNA wash buffer, DNA elution buffer, Human RPL30 positive control primer pair
Specifications
Specifications
| Kit Contents | RPL30 primer, 10x glycine, IP dilution buffer, H3 K4Me3 positive control antibody, Protein A/G magnetic beads, IP wash buffer 1, IP wash buffer 2, IP wash buffer 3, IP wash buffer 4, IP elution buffer, 5M NaCL, Proteinase K, Purification columns, Purification column Tubes, DNA binding buffer, DNA wash buffer, DNA elution buffer, Human RPL30 positive control primer pair |
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Quantity | 30 Immunoprecipitations |
| For Use With (Application) | Chromatin Immunoprecipitation analysis of HeLa nuclear extracts |
| Target | Chromata ChIP |
| Type | ChIP Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.