Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Educational Insights Now You See It, Now You Don't™ See-Through Compost Container
SCI_ED

Catalog No. S02735
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S02735 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02735 Supplier Educational Insights Supplier No. 5095
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Get an up-close view of decomposing objects with the Now You See It, Now You Don't See-Through Compost Containers! Conduct side-by-side experiments in three separate containers: compare three of the same objects in different soils or 3 different objects in the same soil.

  • Get an up-close view of decomposing objects with the Now You See It, Now You Don't See-Through Compost Containers
  • Conduct side-by-side experiments in three separate containers: compare three of the same objects in different soils or 3 different objects in the same soil.
  • Perfect for science fair projects.

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade K to 12
Class Size 1 Class
Product Type Compost Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.