Learn More
Solstice NOWPak™ liner-based bottle system, Accessory, 20L Tilt Dispenser (Stainless)
Catalog No. 6002744 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
60-027-44 Each
1 options
Catalog No. 60-027-44 Supplier Solstice Supplier No. NP523
Add to Cart
Add to Cart
For use with PTFE Pour Spout Closure
  • When using the NP503 PTFE Pour Spout Closure, this metal cradle aids pouring solvent from 20L NOWPak™ containers.
  • Due to overpack size differences, the NP523 Tilt Cradle can be used with HDPE 20L NOWPak™ containers only.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each
WARNING: Reproductive Harm - www.P65Warnings.ca.gov