Learn More
Solstice NOWPak™ liner-based bottle system, Accessory, Cap Adapter Kit - 1/4in. Tubing
Catalog No. 6002940 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
60-029-40 Each
1 options
Catalog No. 60-029-40 Supplier Solstice Supplier No. NP500
Add to Cart
Add to Cart
For direct instrument connection
Cap adapters easily connect NOWPak™ and NOWPak™ II containers directly to analytical HPLC instruments, preparative scale HPLC instruments, GPC instruments and other equipment using pumps or vacuum to supply solvent.
This adapter is suitable for use with high-flow pump or vacuum applications where sparging is not required. It comes with: A cap closure and PTFE adapter with a 4 in. PFA elbow solvent fitting, An FEP diptube with a 40-micron stainless steel frit to prevent drips while changing containers. The 1/4 in. elbow fitting can be replaced with any standard 1/4 in. male NPT fitting for adaptation to other tubing sizes.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each
WARNING: Reproductive Harm - www.P65Warnings.ca.gov