Learn More
Solstice NOWPak™ liner-based bottle system, Accessory, Cap Adapter Kit - 1/8in. Tubing
Catalog No. 6002939 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
60-029-39 Each
1 options
Catalog No. 60-029-39 Supplier Solstice Supplier No. NP514
Add to Cart
Add to Cart
For direct instrument connection
Cap adapters easily connect NOWPak™ and NOWPak™ II containers directly to analytical HPLC instruments, preparative scale HPLC instruments, GPC instruments and other equipment using pumps or vacuum to supply solvent.
This connector is suitable for use with low-flow pump or vacuum applications where sparging is not required. The adapter includes: A cap closure and PTFE adapter with an 8 in. PFA solvent tube fitting, 10' piece of 8 in. FEP solvent tubing with a 40-micron stainless steel frit to prevent drips while changing containers.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each
WARNING: Reproductive Harm - www.P65Warnings.ca.gov