Learn More
Solstice NOWPak™ liner-based bottle system, Accessory, HPLC Sparging Adapter
Catalog No. 6003032 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
60-030-32 Each
1 options
Catalog No. 60-030-32 Supplier Solstice Supplier No. NP512
For direct instrument connection
Cap adapters easily connect NOWPak™ and NOWPak™ II containers directly to analytical HPLC instruments, preparative scale HPLC instruments, GPC instruments and other equipment using pumps or vacuum to supply solvent.
A standard HPLC 4L bottlecap sparging assembly (not included) can be connected directly to the NOWPak™ container for feeding single or multiple HPLC and GPC instruments. This adapter consists of a cap closure and PTFE adapter with a 38 mm thread pattern.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each