Learn More
Solstice NOWPak™ liner-based bottle system, Accessory, Pour Spout Closure
Catalog No. 6003059 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
60-030-59 Each
1 options
Catalog No. 60-030-59 Supplier Solstice Supplier No. NP503
Add to Cart
Add to Cart
For use with NOWPak™ Containers
The PTFE Pour Spout Closure connects to the neck of the NOWPak™ liner for inert, splashless pouring by either lifting or using the NP523 Tilt Cradle. An attached cap allows the container to be resealed after each use without detaching the pour spout.
TRUSTED_SUSTAINABILITY
Specifications
Quantity Each