Learn More
Sigma Aldrich Fine Chemicals Biosciences NOX Inhibitor IV GKT136901
Catalog No. 502898320
Encompass
An orally bioavailable non-toxic pyrazolopyridinedione based compound that acts as a potent and reversible inhibitor of NOX1- and NOX4-containing NADPH oxidase activity (Ki 160 and 165 nM respectively Amplex Red assay) and devoid of antioxidant properties. At higher concentrations affects NOX2 (Ki 1.53 uM). Blocks NOX-mediated ROS (reactive oxygen species) outburst in NOX isozyme expressing PMN cell membranes. Exhibits poor affinity towards xanthine oxidase lipoxygenases-5/12 myeloperoxidase MAO-B iNOS and eNOS several cytochrome P450 isozymes PTP1B and a panel containing representative ion-channels and kinases at 10 181M. functions as a selective scavenger of peroxynitrite over nitric oxide superoxide radical anion and hydroxyl radical.Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided re
Catalog No. 50-289-8320 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 5340320001
An orally bioavailable non-toxic pyrazolopyridinedione based compound that acts as a potent and reversible inhibitor of NOX1- and NOX4-containing NADPH oxidase activity (Ki 160 and 165 nM respectively Amplex Red assay) and devoid of antioxidant properties. At higher concentrations affects NOX2 (Ki 1.53 uM). Blocks NOX-mediated ROS (reactive oxygen species) outburst in NOX isozyme expressing PMN cell membranes. Exhibits poor affinity towards xanthine oxidase lipoxygenases-5/12 myeloperoxidase MAO-B iNOS and eNOS several cytochrome P450 isozymes PTP1B and a panel containing representative ion-channels and kinases at 10 181M. functions as a selective scavenger of peroxynitrite over nitric oxide superoxide radical anion and hydroxyl radical.Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided re