Learn More
Fine Science Tools Noyes Spring Scissors
SDP
Catalog No. NC9740964
Encompass
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 14mm/ Tip Diameter: 0.275mm/ Alloy / Material: Stainless Steel/ Length: 12cm/ Serrated: No
Catalog No. NC9740964 Supplier Fine Science Tools Supplier No. 1501112
Tip Shape: Curved/ Tips: Sharp/ Cutting Edge: 14mm/ Tip Diameter: 0.275mm/ Alloy / Material: Stainless Steel/ Length: 12cm/ Serrated: No