Learn More
Med Vet International Miltex Vantage Noyes Forceps, 6.5in.
Catalog No. 502830282
Encompass_Preferred
The Miltex Vantage V920-142 Noyes Forceps are generally used for ENT procedures. This 6.5in. Noyes Forceps are made of mid-grade stainless steel and may be used are used for removing objects or fluids from the ear canal.
Catalog No. 50-283-0282 Supplier Med Vet International Supplier No. MILV920142
The Miltex Vantage V920-142 Noyes Forceps are generally used for ENT procedures. This 6.5in. Noyes Forceps are made of mid-grade stainless steel and may be used are used for removing objects or fluids from the ear canal.