Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Noyes Micro Dissecting Scissors

Catalog No. 21124916 Shop All Surgical Design Products
Change view
Click to view available options
Tip Style:
Curved
Straight
Color:
Multicolor
Silver
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style Color
21-124-916 Straight Multicolor
21-124-913 Curved Silver
21-124-914 Curved Multicolor
21-124-915 Straight Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-124-916 Supplier Surgical Design™ Supplier No. MSS475STRR
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-precision micro dissecting scissors are designed for fine, controlled cutting in surgical, laboratory, and histology settings.

  • Tip Style: Straight or Curved
  • Color: Silver or Multicolor
  • Fine micro-dissection blades ideal for detailed and delicate cutting tasks
  • Lightweight 4.75 in. design ensures optimal control and maneuverability
  • Precision-ground cutting edges deliver clean, accurate cuts on soft tissues
  • Available in straight or curved tips for versatility across procedures
  • Offered in silver or multicolor finish to suit personal preference or instrument coding
  • Made from premium stainless steel for durability, corrosion resistance, and repeated sterilization

Specifications

Product Type Dissecting Scissors
Color Multicolor
Material Stainless Steel
Tip Style Straight
Length (English) 4.75 in.
Length (Metric) 12 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.