Learn More
Roboz Surgical NOYES SCIS 4.5 SLT CURVE S/S
SDP
Catalog No. NC0441632
Encompass
Noyes Micro Dissecting Spring Scissors; Slightly Curved, Sharp Points; 13mm Cutting Edge; 0.25mm Tip Width; 4-1/2 in. Overall Length
Catalog No. NC0441632 Supplier Roboz Surgical Supplier No. RS5675
Noyes Micro Dissecting Spring Scissors; Slightly Curved, Sharp Points; 13mm Cutting Edge; 0.25mm Tip Width; 4-1/2 in. Overall Length