Learn More
Roboz Surgical NOYES SCISSORS
SDP
Catalog No. NC9466910
Encompass
These micro dissecting spring scissors are designed for delicate surgical procedures, featuring a straight configuration with sharp points. They offer precise control and stability, suitable for fine tissue dissection in laboratory and surgical environments.
  • Straight design with sharp points for precise cutting
  • Overall instrument length of 4 1/2 in.
  • Cutting edge length of 13 mm
  • Spring-action handle for improved maneuverability and control
Catalog No. NC9466910 Supplier Roboz Surgical Supplier No. RS5676
These micro dissecting spring scissors are designed for delicate surgical procedures, featuring a straight configuration with sharp points. They offer precise control and stability, suitable for fine tissue dissection in laboratory and surgical environments.
  • Straight design with sharp points for precise cutting
  • Overall instrument length of 4 1/2 in.
  • Cutting edge length of 13 mm
  • Spring-action handle for improved maneuverability and control