Learn More
Roboz Surgical NOYES SCS 475IN ANG/FLAT 12MM
SDP
Catalog No. 5000574832
Encompass
These micro dissecting spring scissors are designed for delicate surgical procedures requiring precise, fine-tip control. Featuring an angled-on-flat design, this instrument is 4 3/4 in. in length and utilizes 12 mm blades with sharp-on-sharp tips for accurate tissue separation.
  • Designed with an angled-on-flat configuration for improved visibility during microsurgical tasks
  • Constructed with sharp-on-sharp tips for precise cutting performance
  • Features a 12 mm blade length suitable for fine dissection
  • Includes a spring-action handle for ergonomic operation
  • Maintains an overall instrument length of 4 3/4 in.
Catalog No. 50-005-74832 Supplier Roboz Surgical Supplier No. RS5788
These micro dissecting spring scissors are designed for delicate surgical procedures requiring precise, fine-tip control. Featuring an angled-on-flat design, this instrument is 4 3/4 in. in length and utilizes 12 mm blades with sharp-on-sharp tips for accurate tissue separation.
  • Designed with an angled-on-flat configuration for improved visibility during microsurgical tasks
  • Constructed with sharp-on-sharp tips for precise cutting performance
  • Features a 12 mm blade length suitable for fine dissection
  • Includes a spring-action handle for ergonomic operation
  • Maintains an overall instrument length of 4 3/4 in.