Learn More
Fine Science Tools Noyes Spring Scissors, Angled Up
SDP
Catalog No. NC1924642
Encompass
Noyes Spring Scissors| Tips: SharpCutting, Edge: 14mmTip, Diameter: 0.275mm, Length:12cm, Alloy/Material: Stainless Steel, Serrated: No Tip, Shape: Angled Up
Catalog No. NC1924642 Supplier Fine Science Tools Supplier No. 1501312
Noyes Spring Scissors| Tips: SharpCutting, Edge: 14mmTip, Diameter: 0.275mm, Length:12cm, Alloy/Material: Stainless Steel, Serrated: No Tip, Shape: Angled Up