Learn More
BD Cell Analysis 3P BD Biosciences 130 um Nozzle Assembly, For BD FACSAria Sorters, Cell Sorting Applications, Each
Supplier:  BD Cell Analysis 3P 643943
Encompass
The BD Biosciences 130 um Nozzle Assembly is designed for optimal cell sorting with BD FACSAria and FACSymphony systems. It ensures a consistent and reproducible drop profile, catering to diverse particle sizes, enhancing sorting accuracy. Intended for research use only, this nozzle is essential for researchers seeking precision and reliability in cell sorting applications. Its compatibility with leading BD instruments makes it a superior choice for advanced scientific workflows.
Catalog No. NC2400655