Learn More
CELLINK LLC NOZZLE UV SHIELDS
SDP
Catalog No. NC2399757
Encompass
Nozzle shields for light-sensitive and UV-crosslinkable bioinks. Fits over dispensing tip hub. Use with 3 mL cartridges. Package of 10 pieces.
Catalog No. NC2399757 Supplier CELLINK LLC Supplier No. OHR000000102
Nozzle shields for light-sensitive and UV-crosslinkable bioinks. Fits over dispensing tip hub. Use with 3 mL cartridges. Package of 10 pieces.