Learn More
Medchemexpress LLC HY-P1812A 1mg Medchemexpress, NP-12 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1812A1MG
Encompass_Preferred
Medchemexpress, HY-P1812A 1mg NP-12 (TFA) CAS: NP-12 (TFA) is a peptide antagonist of the PD-1 signaling pathway , displays equipotent antagonism toward PD-L1 and PD-L2 in rescue of lymphocyte proliferation and effector functions. NP-12 (TFA) exhibits immune activation, excellent antitumor activity, and potential for better management of immune-related adverse events (irAEs) [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4248