Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NP-40 Surfact-Amps™ Detergent Solution

Catalog No. PI28324 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
6 x 10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI28324 6 x 10 mL
PI85124 50 mL
PI85125 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI28324 Supplier Thermo Scientific™ Supplier No. 28324
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized and supplied as a 10%solution.

Thermo Scientific Surfact-Amps NP-40 Detergent Solution is highly-purified NP-40 detergent stabilized as a 10% solution in your choice of either 10 mL glass ampules or two sizes of HDPE-plastic bottles.

Features of Surfact-Amps NP-40 Detergent Solution:

  • NP-40—a nonionic detergent for use in various protein methods
  • Accurate—precise 10% detergent solutions in ultrapure water
  • Easy-to-use—solutions are simple to dispense and dilute for use
  • Exceptionally pure—less than 1.0μeq/mL peroxides and carbonyls
  • Stable—packaged under inert nitrogen gas in HDPE bottles

This is an easy-to-use 10% (w/v) solution of purified NP-40 detergent for use in routine and high-demand protein research methods and molecular biology techniques. NP-40 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or other solutions intended to extract and solubilize proteins. Surfact-Amps Detergent Solutions (10% w/v) provide unsurpassed purity, quality and stability. Unlike neat detergents, which are extremely viscous, Surfact-Amps 10% Solutions are easy to pipette and accurately dispense. The surfactant solutions are carefully prepared and packaged under nitrogen in glass ampules or non-leaching HDPE bottles, ensuring their stability and eliminating the accumulation of peroxides and degradation products.

Properties of NP-40 Detergent:

  • Molecular Weight: 617 g
  • Detergent Class: Nonionic
  • Aggregation Number: 149
  • Micelle Molecular Weight: 90,000 g
  • Critical Micelle Concentration (CMC): 0.29 mM (0.0179%, w/v)
  • Cloud Point: 80°C
  • Dialyzable: No

Specifications for NP-40 Surfact-Amps Detergent Solution:

  • Visual: Clear, colorless solution, free of particulate matter.
  • Concentration: 10.0±1.0%
  • Oxidants: ≤1.0μeq/mL
  • Carbonyls: ≤1.0μeq/mL
  • Suspended Solids: Residue present must not exceed Residue Reference.

Related Products

  • Surfact-Amps™ Detergent Sampler (Cat. No. 28340)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Detergent
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C.
Form Liquid
Product Line Surfact-Amps
Reagent Type Detergent Solution
Quantity 6 x 10 mL
Format Glass Ampule

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.