Learn More
Boston Bioproducts Inc NP-40 Lysis Buffer (2X) - 250ml
Catalog No. NC2249956
Encompass
The 2X NP-40 Lysis Buffer is a solubilizing agent used to prepare cell extracts for immunoassays and blotting techniques. As one of the most popular lysis buffers on the market, the NP-40 lysis buffers can support a variety of laboratory workflows. Relying on the non-ionic detergent NP-40 as its primary solubilizing agent, this Lysis buffer is weaker than a RIPA buffer, and can permeabilize cell membranes, partially open the nucleus without fully disrupting it, and lyse mitochondiral membranes.
This item is not returnable. View return policy
Catalog No. NC2249956 Supplier Boston Bioproducts Inc Supplier No. BP119X250ML
The 2X NP-40 Lysis Buffer is a solubilizing agent used to prepare cell extracts for immunoassays and blotting techniques. As one of the most popular lysis buffers on the market, the NP-40 lysis buffers can support a variety of laboratory workflows. Relying on the non-ionic detergent NP-40 as its primary solubilizing agent, this Lysis buffer is weaker than a RIPA buffer, and can permeabilize cell membranes, partially open the nucleus without fully disrupting it, and lyse mitochondiral membranes.