Learn More
Bioworld NP-40 Lysis Buffer (High Salt), 250 mL
Catalog No. 50253041
Encompass_Preferred
NP-40 Lysis Buffer (High Salt) Lysis buffer for extracting proteins from cells. Technical Information:1X concentrationTris-HCL, pH 8.0 (50mM)NaCl (500mM)NP-40 (1%)EDTA (5mM)
Catalog No. 50-253-041 Supplier Bioworld Supplier No. 220400441
NP-40 Lysis Buffer (High Salt) Lysis buffer for extracting proteins from cells. Technical Information:1X concentrationTris-HCL, pH 8.0 (50mM)NaCl (500mM)NP-40 (1%)EDTA (5mM)