Learn More
Bioworld NP-40 Lysis Buffer (Low Salt), 250 mL
Catalog No. 50253043
Encompass_Preferred
NP-40 Lysis BufferLysis buffer for extracting proteins from cells Low saltSpecifications:pH: 8.01X concentrationContains: 50 mM Tris-HCL, 5mM EDTA, 150mM NaCl and 1% NP-40
Catalog No. 50-253-043 Supplier Bioworld Supplier No. 220400451
NP-40 Lysis BufferLysis buffer for extracting proteins from cells Low saltSpecifications:pH: 8.01X concentrationContains: 50 mM Tris-HCL, 5mM EDTA, 150mM NaCl and 1% NP-40