Learn More
Bioworld NP-40 Lysis Buffer (No Salt), 500 mL
Catalog No. 50253046
Encompass_Preferred
NP-40 Lysis Buffer Lysis buffer for extracting proteins from cells. Technical Information:1X concentrationTris-HCL, pH 8.0 (50mM)NP-40 (1%)EDTA (5mM)
Catalog No. 50-253-046 Supplier Bioworld Supplier No. 220400462
NP-40 Lysis Buffer Lysis buffer for extracting proteins from cells. Technical Information:1X concentrationTris-HCL, pH 8.0 (50mM)NP-40 (1%)EDTA (5mM)