Learn More
Aviva Systems Biology Corp NP Antibody - middle region (ARP48474_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48474_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NP. Alternate names: MGC117396, MGC125915, MGC125916, PNP, PRO1837, PUNP, NPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59837