Learn More
Medchemexpress LLC HY-125506 5mg Medchemexpress, NP-G2-044 CAS:1807454-59-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1255065MG
Encompass_Preferred
Medchemexpress, HY-125506 5mg NP-G2-044 CAS:1807454-59-6 NP-G2-044 is a potent, orally active fascin inhibitor, with an IC50 of ~2 μM. NP-G2-044 blocks tumor metastasis and increases antitumor immune response[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0027