Learn More
Medchemexpress LLC Np-OVAL (Ovalbumin) | 5 MG
SDP
Catalog No. 5000426064
Encompass_Preferred
NP-OVAL (Ovalbumin) is a complex formed by 4-Hydroxy-3-nitrophenylacetyl (NP, a hapten) with Ovalbumin (OVAL, a carrier protein). NP-OVAL (Ovalbumin) can induce specific immune responses and NP-specific antibodies production. NP-OVAL (Ovalbumin) can accelerate the construction of multiple myeloma models.
  • Can induce specific immune responses and NP-specific antibodies production.
  • Can accelerate the construction of multiple myeloma models.
Catalog No. 50-004-26064 Supplier Medchemexpress LLC Supplier No. HYNP1385MG
NP-OVAL (Ovalbumin) is a complex formed by 4-Hydroxy-3-nitrophenylacetyl (NP, a hapten) with Ovalbumin (OVAL, a carrier protein). NP-OVAL (Ovalbumin) can induce specific immune responses and NP-specific antibodies production. NP-OVAL (Ovalbumin) can accelerate the construction of multiple myeloma models.
  • Can induce specific immune responses and NP-specific antibodies production.
  • Can accelerate the construction of multiple myeloma models.