Learn More
Trajan Scientific NP0.5BN-C/F-0.47 REPAIR KIT
SDP
Catalog No. 502530269
Encompass_Preferred
Trajan Replacement Needles for Thermo Scientific GC Syringes, Volume: 0.5 uL, Diameter Outer Needle: 0.47 mm, Needle Gauge: 26 G, Length Needle: 50 mm, Needle Point Style: Cone, Diameter Inner Needle: 0.155 mm, GC Autosampler Syringe Replacement Needle
Catalog No. 50-253-0269 Supplier Trajan Scientific Supplier No. 033770
Trajan Replacement Needles for Thermo Scientific GC Syringes, Volume: 0.5 uL, Diameter Outer Needle: 0.47 mm, Needle Gauge: 26 G, Length Needle: 50 mm, Needle Point Style: Cone, Diameter Inner Needle: 0.155 mm, GC Autosampler Syringe Replacement Needle