Learn More
Apexbio Technology LLC NP118809 41332-24-5 200mg
SDP
Catalog No. 5000093750
Encompass_Preferred
NP118809 (CAS 41332-24-5) is a potent and selective blocker of N-type voltage-gated calcium channels exhibiting an IC50 of 0 11 M in vitro It demonstrates significant selectivity over L-type calcium channels with over 100-fold preference (L-type IC50 12 2 M) In vivo studies in rats indicate that NP118809 is orally bioavailable (mean 30%) with a Tmax of 2 3 hours and a half-life of 2 1 hours following intravenous or oral administration The compound exhibits wide tissue distribution and has been shown to produce analgesic effects in animal pain models NP118809 is useful for research into N-type calcium channel function and modulation particularly in studies of pain signaling and neuropharmacology
Catalog No. 50-000-93750 Supplier Apexbio Technology LLC Supplier No. A3666200
NP118809 (CAS 41332-24-5) is a potent and selective blocker of N-type voltage-gated calcium channels exhibiting an IC50 of 0 11 M in vitro It demonstrates significant selectivity over L-type calcium channels with over 100-fold preference (L-type IC50 12 2 M) In vivo studies in rats indicate that NP118809 is orally bioavailable (mean 30%) with a Tmax of 2 3 hours and a half-life of 2 1 hours following intravenous or oral administration The compound exhibits wide tissue distribution and has been shown to produce analgesic effects in animal pain models NP118809 is useful for research into N-type calcium channel function and modulation particularly in studies of pain signaling and neuropharmacology