Learn More
Medchemexpress LLC NP3-562 | 2409825-32-5 | 98.2% | 437.94 | 50 MG
SDP
Catalog No. 5000395000
Encompass_Preferred
NP3-562 is an orally active NLRP3 Inhibitor that blocks IL-1β release in stimulated cells and in vivo models. It is suitable for acute peritonitis research.
  • Orally active NLRP3 inhibitor
  • Inhibits IL-1β release
  • Used for acute peritonitis studies
  • Demonstrates brain exposure
Catalog No. 50-003-95000 Supplier Medchemexpress LLC Supplier No. HY16315950MG
NP3-562 is an orally active NLRP3 Inhibitor that blocks IL-1β release in stimulated cells and in vivo models. It is suitable for acute peritonitis research.
  • Orally active NLRP3 inhibitor
  • Inhibits IL-1β release
  • Used for acute peritonitis studies
  • Demonstrates brain exposure