Learn More
Cayman Chemical Np63N-Term RabbIt Monclo 100uL
SDP
Catalog No. 502948437
Encompass_Preferred
An mPGES-1 inhibitor (IC50s 117 and 60 nM for the human and mouse enzymes respectively) inhibits LPS-induced PGE2 production in mouse peritoneal cells (IC50 31 nM) reduces carrageenan-induced paw edema in rats at 25 mg/kg
Catalog No. 50-294-8437 Supplier Cayman Chemical Supplier No. 32316100 UL
An mPGES-1 inhibitor (IC50s 117 and 60 nM for the human and mouse enzymes respectively) inhibits LPS-induced PGE2 production in mouse peritoneal cells (IC50 31 nM) reduces carrageenan-induced paw edema in rats at 25 mg/kg