Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NP69 Human Nasopharyngeal Epithelial Cell Line

Catalog No. SCC197MI Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Cells:
≥ 1 x 106 Cells/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Cells
SCC197MI ≥ 1 x 106 Cells/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC197MI Supplier MilliporeSigma™ Supplier No. SCC197
Add to Cart
Add to Cart

NP69 cell line used in studies of mechanisms of viral-associated tumorigenesis

  • Retains many of the characteristics of normal nasopharyngeal cells, including keratin profiles and response to TGF-beta, while possessing many of the genetic signatures observed in NPC
  • NP69 cells are non-tumorigenic and demonstrate anchorage-dependent growth
  • Uses:
    • For studies on mechanisms of viral-associated tumorigenesis
  • Contains genetically modified organisms (GMO)

    TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in liquid nitrogen, Cells can be cultured for at least 10 passages after initial thawing without significantly affecting cell marker expression and functionality
    Cell Type Epithelial Cells
    For Use With (Application) Cell Culture
    No. of Cells ≥ 1 x 106 Cells/Vial

    For research purposes only

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.