Learn More
Research Products International Corp NPA (N-1-Naphthylphtalamic Acid), 250 Milligrams
SDP
Catalog No. 50213562
Encompass_Preferred
Non-competitive transport inhibitor of auxin.
Catalog No. 50-213-562 Supplier Research Products International Corp Supplier No. N420600.25
Non-competitive transport inhibitor of auxin.