Learn More
Aviva Systems Biology Corp NPAL2 Antibody - N-terminal region (ARP49863_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49863_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NPAL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NPAL2. Alternate names: FLJ13955, NPAL2ApplicationWB. Species reactivity: Human, Rabbit, Rat
Catalog No. 50-014-60205