Learn More
Aviva Systems Biology Corp Npas1 Antibody - middle region (ARP36951_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36951_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Npas1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Npas1. Alternate names: MOP5, bHLHe11ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-549